Product Details

SNP ID
rs114626579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62956895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTGGAACAAGAAGGAGGCCGGC[A/T]TCGTGCTCAGCAGCTTCTTCTGGGG
Phenotype
MIM: 611625 MIM: 612107
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GID8 PubMed Links

Gene Details

Gene
GID8
Gene Name
GID complex subunit 8 homolog
There are no transcripts associated with this gene.

Gene
SLC17A9
Gene Name
solute carrier family 17 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302643.1 436 Missense Mutation ATC,TTC I58F NP_001289572.1
NM_022082.3 436 Missense Mutation ATC,TTC I64F NP_071365.3
XM_011528978.2 436 UTR 5 XP_011527280.1

View Full Product Details