Product Details

SNP ID
rs112732285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63490632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCAGAGCCTCGTGGTGCATCTC[C/T]ACTGACTTCACCTCAGTGGTGATGT
Phenotype
MIM: 602959
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEF1A2 PubMed Links

Gene Details

Gene
EEF1A2
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001958.3 1205 Silent Mutation GTA,GTG V292V NP_001949.1

View Full Product Details