Product Details

SNP ID
rs117222808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43460018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTAAGATTTCCGAGTCTGACC[A/C]ATCTTGTCTTTCAGGATTTTATGCG
Phenotype
MIM: 601944
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRSF6 PubMed Links

Gene Details

Gene
SRSF6
Gene Name
serine and arginine rich splicing factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006275.5 Intron NP_006266.2

View Full Product Details