Product Details

SNP ID
rs117018348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44969981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCATTACATTTGTGAATCCATTT[C/T]ACAAATATATTTTGACCACCTCTTG
Phenotype
MIM: 604965 MIM: 616049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK4 PubMed Links

Gene Details

Gene
STK4
Gene Name
serine/threonine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006282.3 Intron NP_006273.1
XM_005260530.2 Intron XP_005260587.1
XM_005260531.3 Intron XP_005260588.1
XM_005260532.3 Intron XP_005260589.1
XM_005260533.2 Intron XP_005260590.1
XM_011529018.2 Intron XP_011527320.1
XM_011529020.2 Intron XP_011527322.1
XM_017028029.1 Intron XP_016883518.1
XM_017028030.1 Intron XP_016883519.1
XM_017028031.1 Intron XP_016883520.1
XM_017028032.1 Intron XP_016883521.1
XM_017028033.1 Intron XP_016883522.1
Gene
STK4-AS1
Gene Name
STK4 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TOMM34
Gene Name
translocase of outer mitochondrial membrane 34
There are no transcripts associated with this gene.

View Full Product Details