Product Details

SNP ID
rs116380320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32631639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCAGGAGGTAAAGAGGAATTAC[A/G]TGGCCTTTTATTCACTACATTTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf203 PubMed Links

Gene Details

Gene
C20orf203
Gene Name
chromosome 20 open reading frame 203
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182584.2 4779 UTR 3 NP_872390.2

View Full Product Details