Product Details

SNP ID
rs116077790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45950068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCCGTGACTGGACTGGAGTGC[A/G]GCTGGGCAGAGAGGAGAGGATGCTC
Phenotype
MIM: 610827
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCIF1 PubMed Links

Gene Details

Gene
PCIF1
Gene Name
PDX1 C-terminal inhibiting factor 1
There are no transcripts associated with this gene.

Gene
ZNF335
Gene Name
zinc finger protein 335
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022095.3 3383 Silent Mutation NP_071378.1
XM_005260504.4 3383 Silent Mutation XP_005260561.1
XM_011528979.2 3383 Intron XP_011527281.1
XM_017028012.1 3383 Silent Mutation XP_016883501.1

View Full Product Details