Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001281457.1 | 2215 | UTR 3 | NP_001268386.1 | ||
NM_001281458.1 | 2215 | UTR 3 | NP_001268387.1 | ||
NM_001281459.1 | 2215 | UTR 3 | NP_001268388.1 | ||
NM_001281460.1 | 2215 | UTR 3 | NP_001268389.1 | ||
NM_015945.11 | 2215 | UTR 3 | NP_057029.8 | ||
NM_173073.3 | 2215 | UTR 3 | NP_775096.1 | ||
NM_173179.3 | 2215 | UTR 3 | NP_775271.1 | ||
XM_011528831.1 | 2215 | Intron | XP_011527133.1 | ||
XM_011528832.1 | 2215 | Intron | XP_011527134.1 | ||
XM_011528833.1 | 2215 | Intron | XP_011527135.1 | ||
XM_011528834.1 | 2215 | Intron | XP_011527136.1 | ||
XM_011528835.1 | 2215 | Intron | XP_011527137.1 | ||
XM_011528836.1 | 2215 | Intron | XP_011527138.1 | ||
XM_011528837.1 | 2215 | Intron | XP_011527139.1 | ||
XM_011528838.1 | 2215 | Intron | XP_011527140.1 | ||
XM_017027856.1 | 2215 | Intron | XP_016883345.1 | ||
XM_017027857.1 | 2215 | Intron | XP_016883346.1 | ||
XM_017027858.1 | 2215 | Intron | XP_016883347.1 | ||
XM_017027859.1 | 2215 | Intron | XP_016883348.1 | ||
XM_017027860.1 | 2215 | Intron | XP_016883349.1 | ||
XM_017027861.1 | 2215 | Intron | XP_016883350.1 | ||
XM_017027862.1 | 2215 | Intron | XP_016883351.1 |