Product Details

SNP ID
rs113814875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:48922468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGCCCGTTGGAGAATGTTTTC[C/T]TTTGGTTTGGGGATCTCCCATAGCT
Phenotype
MIM: 605371
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARFGEF2 PubMed Links

Gene Details

Gene
ARFGEF2
Gene Name
ADP ribosylation factor guanine nucleotide exchange factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006420.2 Intron NP_006411.2
XM_005260252.3 Intron XP_005260309.1

View Full Product Details