Product Details
- SNP ID
-
rs112210429
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:49633436 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATATGCACAAGGTCACATTTGGTT[C/T]CTGTTTTTTTTTTTAACATGACAAT
- Phenotype
-
MIM: 604016
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
B4GALT5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11471493] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- B4GALT5
- Gene Name
- beta-1,4-galactosyltransferase 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004776.3 |
4237 |
UTR 3 |
|
|
NP_004767.1 |
View Full Product Details