Product Details

SNP ID
rs116061842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33287669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGTCCACAGTTGGCAATCTGA[C/T]GCTAGTGGGTGGAGAACTACCTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPIFB1 PubMed Links

Gene Details

Gene
BPIFB1
Gene Name
BPI fold containing family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033197.2 Intron NP_149974.2

View Full Product Details