Product Details

SNP ID
rs114995219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56422797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCACAGCATGTGCCATTCTCCA[C/T]GATTATTTTTAGAGGTCTTAAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASS4 PubMed Links

Gene Details

Gene
CASS4
Gene Name
Cas scaffolding protein family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164114.1 Intron NP_001157586.1
NM_001164115.1 Intron NP_001157587.1
NM_001164116.1 Intron NP_001157588.1
NM_020356.3 Intron NP_065089.2
XM_006723831.3 Intron XP_006723894.1

View Full Product Details