Product Details

SNP ID
rs116115328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34560895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGCACGAGCATGGCCTCTGTGC[C/T]ATCCTTGGCGGTCAAGTAGAGGCCA
Phenotype
MIM: 601242 MIM: 608528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1LC3A PubMed Links

Gene Details

Gene
MAP1LC3A
Gene Name
microtubule associated protein 1 light chain 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032514.3 1479 Intron NP_115903.1
NM_181509.2 1479 Intron NP_852610.1
XM_011529083.2 1479 Intron XP_011527385.1
XM_011529084.2 1479 Intron XP_011527386.1
XM_011529085.2 1479 Intron XP_011527387.1
Gene
PIGU
Gene Name
phosphatidylinositol glycan anchor biosynthesis class U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080476.4 1479 Missense Mutation AGC,GGC S427G NP_536724.1
XM_011528542.2 1479 Missense Mutation AGC,GGC S211G XP_011526844.1
XM_017027664.1 1479 Missense Mutation AGC,GGC S379G XP_016883153.1
XM_017027665.1 1479 Intron XP_016883154.1

View Full Product Details