Product Details

SNP ID
rs121909350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35434282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCCAGTGGGGCCCGCCGTTTT[T/C]GCCGCTGGCTGAACAGGTACTCATA
Phenotype
MIM: 601146
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GDF5 PubMed Links

Gene Details

Gene
GDF5
Gene Name
growth differentiation factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000557.4 1395 Missense Mutation CAA,CGA Q378R NP_000548.2
NM_001319138.1 1395 Missense Mutation CAA,CGA Q378R NP_001306067.1
XM_011529075.2 1395 Missense Mutation CAA,CGA Q378R XP_011527377.1

View Full Product Details