Product Details
- SNP ID
-
rs118047004
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31306377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGATAAAAGGAAAAATTTGTTTGG[A/G]TTGTCCTAATAGAAAATGTTGTCCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB116
PubMed Links
Gene Details
- Gene
- DEFB116
- Gene Name
- defensin beta 116
View Full Product Details