Product Details

SNP ID
rs117384548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1310069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAGTCAAGGCAAGGAAAAAGCT[C/G]TCACAAAGAACGTAGCTCTGTTCTC
Phenotype
MIM: 604942
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FKBP1A-SDCBP2 PubMed Links

Gene Details

Gene
FKBP1A-SDCBP2
Gene Name
FKBP1A-SDCBP2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SDCBP2
Gene Name
syndecan binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199784.1 1421 UTR 3 NP_001186713.1
NM_015685.5 1421 UTR 3 NP_056500.2
NM_080489.4 1421 UTR 3 NP_536737.3
Gene
SNPH
Gene Name
syntaphilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318234.1 1421 Intron NP_001305163.1
NM_014723.3 1421 Intron NP_055538.2
XM_005260889.3 1421 Intron XP_005260946.1
XM_011529402.2 1421 Intron XP_011527704.1
XM_011529403.2 1421 Intron XP_011527705.1
XM_011529404.2 1421 Intron XP_011527706.1

View Full Product Details