Product Details

SNP ID
rs118149759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13716046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAATGGGAGCATATGTGAAACT[A/G]ACTCAAACAACAGAAGCTACAGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESF1 PubMed Links

Gene Details

Gene
ESF1
Gene Name
ESF1 nucleolar pre-rRNA processing protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276380.1 Intron NP_001263309.1
NM_016649.3 Intron NP_057733.2
XM_017027874.1 Intron XP_016883363.1

View Full Product Details