Product Details

SNP ID
rs116865999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:17615954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAACTCGGCCGTGAGCTTCTGC[C/T]GCTGTGTCTGCTCATCCTCCAGGAT
Phenotype
MIM: 609114 MIM: 601418
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSTN PubMed Links

Gene Details

Gene
DSTN
Gene Name
destrin, actin depolymerizing factor
There are no transcripts associated with this gene.

Gene
RRBP1
Gene Name
ribosome binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042576.1 2978 Missense Mutation CAG,CGG Q875R NP_001036041.1
NM_004587.2 2978 Missense Mutation CAG,CGG Q875R NP_004578.2

View Full Product Details