Product Details

SNP ID
rs111925487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:271129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATCAGGGTCTGAATGGAGATCC[A/G]GTCCTGGAAGTAAATGATCCAAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf96 PubMed Links

Gene Details

Gene
C20orf96
Gene Name
chromosome 20 open reading frame 96
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080571.1 1268 UTR 3 NP_542138.1
NM_153269.2 1268 UTR 3 NP_695001.2
XM_006723543.3 1268 UTR 3 XP_006723606.1

View Full Product Details