Product Details

SNP ID
rs114569807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:326656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTGAGGAAGGTGAAGAGGAGAC[A/G]GTGGCGTCGGGGGAGGAGTCGCTGG
Phenotype
MIM: 601947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRSN2-AS1 PubMed Links

Gene Details

Gene
NRSN2-AS1
Gene Name
NRSN2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SOX12
Gene Name
SRY-box 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006943.3 1086 Silent Mutation ACA,ACG T244T NP_008874.2

View Full Product Details