Product Details

SNP ID
rs112349117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:24963439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTTGAGGCCGCAGGACAGGGCC[A/G]CAGCAGCAAGCTCTCTGGGTAGTGC
Phenotype
MIM: 615884 MIM: 603253
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APMAP PubMed Links

Gene Details

Gene
APMAP
Gene Name
adipocyte plasma membrane associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020531.2 1726 UTR 3 NP_065392.1
XM_005260763.3 1726 UTR 3 XP_005260820.1
Gene
CST7
Gene Name
cystatin F
There are no transcripts associated with this gene.

View Full Product Details