Product Details

SNP ID
rs111964495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3889229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCACCACCCTCTCCCCGCC[C/T]CGTCACGATAGCCTCTCATTGGACG
Phenotype
MIM: 606157
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PANK2 PubMed Links

Gene Details

Gene
PANK2
Gene Name
pantothenate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324191.1 296 UTR 5 NP_001311120.1
NM_001324192.1 296 Silent Mutation CCC,CCT P43P NP_001311121.1
NM_001324193.1 296 Intron NP_001311122.1
NM_024960.5 296 Intron NP_079236.3
NM_153638.3 296 Silent Mutation CCC,CCT P43P NP_705902.2
NM_153640.3 296 Intron NP_705904.1
XM_005260835.3 296 Intron XP_005260892.1
XM_005260836.4 296 Intron XP_005260893.3
XM_011529364.2 296 Silent Mutation CCC,CCT P43P XP_011527666.1
XM_011529365.2 296 Silent Mutation CCC,CCT P43P XP_011527667.1
XM_017028077.1 296 Intron XP_016883566.1
XM_017028078.1 296 Intron XP_016883567.1
XM_017028079.1 296 Intron XP_016883568.1

View Full Product Details