Product Details

SNP ID
rs112617006
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:6041709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAATCTCGCCCTCGGAAAGACT[C/T]GGGTTGGGGGCTTGGGGTGTGGCGT
Phenotype
MIM: 608188
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CRLS1 PubMed Links

Gene Details

Gene
CRLS1
Gene Name
cardiolipin synthase 1
There are no transcripts associated with this gene.

Gene
LRRN4
Gene Name
leucine rich repeat neuronal 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152611.4 772 Silent Mutation NP_689824.2
XM_011529183.2 772 Silent Mutation XP_011527485.1

View Full Product Details