Product Details

SNP ID
rs116645811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25587758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGAGGTAGTGAAGAATGACTGC[A/G]TAGTAAATAGAAGGAGAATGGGAAA
Phenotype
MIM: 611845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00515 PubMed Links

Gene Details

Gene
LINC00515
Gene Name
long intergenic non-protein coding RNA 515
There are no transcripts associated with this gene.

Gene
MRPL39
Gene Name
mitochondrial ribosomal protein L39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017446.3 1007 Intron NP_059142.2
NM_080794.3 1007 Missense Mutation ACG,ATG T334M NP_542984.2
XM_006724026.3 1007 Missense Mutation ACG,ATG T334M XP_006724089.1
XM_011529651.2 1007 Intron XP_011527953.1

View Full Product Details