Product Details
- SNP ID
-
rs112325952
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:39600451 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATTTAGAGATTGGAAGATCTAGG[C/T]GTTAAGCCATTTTCTAATGCTGTAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
B3GALT5-AS1
PubMed Links
Gene Details
- Gene
- B3GALT5-AS1
- Gene Name
- B3GALT5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details