Product Details

SNP ID
rs116164005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:18269435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAGCTTACAATAAATAGTTTCT[A/G]TTGGTTGGGAAAATACTAAGGTTAT
Phenotype
MIM: 607247 MIM: 606635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHODL PubMed Links

Gene Details

Gene
CHODL
Gene Name
chondrolectin
There are no transcripts associated with this gene.

Gene
TMPRSS15
Gene Name
transmembrane protease, serine 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002772.2 3789 UTR 3 NP_002763.2
XM_011529654.1 3789 UTR 3 XP_011527956.1
XM_011529655.1 3789 UTR 3 XP_011527957.1
XM_011529656.2 3789 UTR 3 XP_011527958.1
XM_011529657.1 3789 UTR 3 XP_011527959.1
XM_011529658.1 3789 UTR 3 XP_011527960.1
XM_011529659.1 3789 UTR 3 XP_011527961.1

View Full Product Details