Product Details

SNP ID
rs116737357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44329459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTGTGGCATCCTCATAAGACTT[A/G]CGACTCTTCCCAGAGGAAAGAAAAC
Phenotype
MIM: 603191 MIM: 171860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C21orf2 PubMed Links

Gene Details

Gene
C21orf2
Gene Name
chromosome 21 open reading frame 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271440.1 1623 UTR 3 NP_001258369.1
NM_001271441.1 1623 UTR 3 NP_001258370.1
NM_001271442.1 1623 UTR 3 NP_001258371.1
NM_004928.2 1623 UTR 3 NP_004919.1
XM_006724051.3 1623 UTR 3 XP_006724114.1
XM_006724053.3 1623 UTR 3 XP_006724116.1
XM_017028470.1 1623 UTR 3 XP_016883959.1
XM_017028471.1 1623 UTR 3 XP_016883960.1
XM_017028472.1 1623 UTR 3 XP_016883961.1
Gene
PFKL
Gene Name
phosphofructokinase, liver type
There are no transcripts associated with this gene.

View Full Product Details