Product Details

SNP ID
rs117899230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41888807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTGCCCTCCACAATCTATTAGA[A/G]CATATTATTTCACTTATAATGTTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD2 PubMed Links

Gene Details

Gene
C2CD2
Gene Name
C2 calcium dependent domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015500.1 1886 UTR 3 NP_056315.1
NM_199050.2 1886 UTR 3 NP_950251.1
XM_005261109.4 1886 UTR 3 XP_005261166.1
XM_011529522.1 1886 UTR 3 XP_011527824.1
XM_011529523.2 1886 UTR 3 XP_011527825.1
XM_011529524.2 1886 Intron XP_011527826.1
XM_017028315.1 1886 UTR 3 XP_016883804.1
Gene
PRDM15
Gene Name
PR domain 15
There are no transcripts associated with this gene.

View Full Product Details