Product Details

SNP ID
rs117385282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42476033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCAGAGCCTGGGCCTCCTCCC[C/T]GGGTTCTCCTGCACCTGAGATAAAA
Phenotype
MIM: 609314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSPH1 PubMed Links

Gene Details

Gene
RSPH1
Gene Name
radial spoke head 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286506.1 910 Missense Mutation AGG,GGG R210G NP_001273435.1
NM_080860.3 910 Missense Mutation AGG,GGG R248G NP_543136.1
XM_005261208.2 910 Missense Mutation AGG,GGG R179G XP_005261265.1
XM_011529786.1 910 Missense Mutation AGG,GGG R224G XP_011528088.1

View Full Product Details