Product Details

SNP ID
rs117750028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19447103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGCAACATCACCTCCTGCGTA[A/G]GCCAGCTGATGACCCTAACACCGGA
Phenotype
MIM: 601754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166242.1 Intron NP_001159714.1
NM_173793.4 Intron NP_776154.3
Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
There are no transcripts associated with this gene.

View Full Product Details