Product Details
- SNP ID
-
rs118063473
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:19950544 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACATGTTTCCAGTGAGCTCAGCT[G/T]GTTGCTTTTGTGGGTCCCTGAGGCA
- Phenotype
-
MIM: 116790
MIM: 606448
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
COMT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141294168] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COMT
- Gene Name
- catechol-O-methyltransferase
- Gene
- TXNRD2
- Gene Name
- thioredoxin reductase 2
There are no transcripts associated with this gene.
View Full Product Details