Product Details
- SNP ID
-
rs111408637
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:20242038 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCGTTGCCCGGCGGGCTGTCACC[A/G]GGCGGCACGCGTCCCTTCAGCGCAT
- Phenotype
-
MIM: 605566
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1286
PubMed Links
Gene Details
- Gene
- MIR1286
- Gene Name
- microRNA 1286
There are no transcripts associated with this gene.
- Gene
- RTN4R
- Gene Name
- reticulon 4 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023004.5 |
1296 |
Silent Mutation |
CCC,CCT |
P365P |
NP_075380.1 |
View Full Product Details