Product Details

SNP ID
rs115546021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30490169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCGTCTCCTCAGAGGCCTTGT[C/T]GGGAAGCAGCACCTCCACAGTGTAG
Phenotype
MIM: 612825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDC4P PubMed Links

Gene Details

Gene
SDC4P
Gene Name
syndecan 4 pseudogene
There are no transcripts associated with this gene.

Gene
SEC14L4
Gene Name
SEC14 like lipid binding 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161368.1 1061 Intron NP_001154840.1
NM_174977.3 1061 Missense Mutation NP_777637.1
XM_006724235.2 1061 Missense Mutation XP_006724298.1
XM_011530164.2 1061 Missense Mutation XP_011528466.1
XM_017028787.1 1061 Missense Mutation XP_016884276.1
XM_017028788.1 1061 Missense Mutation XP_016884277.1
XM_017028789.1 1061 Missense Mutation XP_016884278.1
XM_017028790.1 1061 Intron XP_016884279.1
XM_017028791.1 1061 Missense Mutation XP_016884280.1

View Full Product Details