Product Details

SNP ID
rs117533839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32810238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTTCAGAGGGGCCCCAGAGAAC[A/C]TTTCCTGACCCATTTAGAGGCCATC
Phenotype
MIM: 602705 MIM: 188826
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SYN3 PubMed Links

Gene Details

Gene
SYN3
Gene Name
synapsin III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135774.1 Intron NP_001129246.1
NM_003490.3 Intron NP_003481.3
NM_133633.2 Intron NP_598344.2
XM_011530405.2 Intron XP_011528707.1
XM_011530406.2 Intron XP_011528708.1
XM_011530407.2 Intron XP_011528709.1
XM_011530408.1 Intron XP_011528710.1
XM_011530410.2 Intron XP_011528712.1
XM_017028960.1 Intron XP_016884449.1
XM_017028961.1 Intron XP_016884450.1
XM_017028962.1 Intron XP_016884451.1
XM_017028963.1 Intron XP_016884452.1
XM_017028964.1 Intron XP_016884453.1
XM_017028965.1 Intron XP_016884454.1
Gene
TIMP3
Gene Name
TIMP metallopeptidase inhibitor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000362.4 Intron NP_000353.1

View Full Product Details