Product Details

SNP ID
rs114305138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21991738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGCAGATCTCCAGGGAATGTA[A/G]TGGGACCCACAGACAGCTGAGGAGA
Phenotype
MIM: 603582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRAMENP PubMed Links

Gene Details

Gene
PRAMENP
Gene Name
PRAME N-terminal-like, pseudogene
There are no transcripts associated with this gene.

Gene
TOP3B
Gene Name
topoisomerase (DNA) III beta
There are no transcripts associated with this gene.

View Full Product Details