Product Details

SNP ID
rs115490807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38081063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAGCCCATTGGCCAGCAGGGC[C/T]AGGCTGAACAGATACGGGACGTGCG
Phenotype
MIM: 605926 MIM: 610409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAIAP2L2 PubMed Links

Gene Details

Gene
BAIAP2L2
Gene Name
BAI1 associated protein 2 like 2
There are no transcripts associated with this gene.

Gene
PICK1
Gene Name
protein interacting with PRKCA 1
There are no transcripts associated with this gene.

Gene
SLC16A8
Gene Name
solute carrier family 16 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013356.2 1160 Silent Mutation NP_037488.2
XM_017028685.1 1160 Silent Mutation XP_016884174.1

View Full Product Details