Product Details

SNP ID
rs115727092
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39225076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCTTATCCCAAGTTCTTGGAGT[C/T]AAGGGAAGAAGATGGCGATGGAGTT
Phenotype
MIM: 190040
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PDGFB PubMed Links

Gene Details

Gene
PDGFB
Gene Name
platelet derived growth factor subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002608.3 2084 UTR 3 NP_002599.1
NM_033016.3 2084 UTR 3 NP_148937.1

View Full Product Details