Product Details

SNP ID
rs114786489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39225814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGGGGCCGGCGGACTCGCACC[A/G]TCCGAATGGTCACCCGAGTTTGGGG
Phenotype
MIM: 190040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDGFB PubMed Links

Gene Details

Gene
PDGFB
Gene Name
platelet derived growth factor subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002608.3 1727 Missense Mutation NP_002599.1
NM_033016.3 1727 Missense Mutation NP_148937.1

View Full Product Details