Product Details
- SNP ID
-
rs115698459
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:39359755 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTCTGTGACCCTTTCCACACCT[A/G]GCCTCCTCTTCCCGCATGGTCTTGT
- Phenotype
-
MIM: 603925
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SYNGR1
PubMed Links
Gene Details
- Gene
- SYNGR1
- Gene Name
- synaptogyrin 1
View Full Product Details