Product Details

SNP ID
rs111399232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22196139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACTGTGATCTATGAGGATAA[C/T]CAAAGACCCTCTGGGGTCCCTGATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGLV6-57 PubMed Links

Gene Details

Gene
IGLV6-57
Gene Name
immunoglobulin lambda variable 6-57
There are no transcripts associated with this gene.

View Full Product Details