Product Details

SNP ID
rs117014665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40005236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTGAGCCCTGCCTCGCTCTGT[C/T]CATGTACTTGAGAGCTCAAGGCTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM83F PubMed Links

Gene Details

Gene
FAM83F
Gene Name
family with sequence similarity 83 member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138435.2 Intron NP_612444.2

View Full Product Details