Product Details

SNP ID
rs117929685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22548039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCTTTAGTGTCCAAGTATGCA[G/T]AATGAAGCATTTGATATGTGCACCC
Phenotype
MIM: 606021
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRAME PubMed Links

Gene Details

Gene
PRAME
Gene Name
preferentially expressed antigen in melanoma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291715.1 2083 UTR 3 NP_001278644.1
NM_001291716.1 2083 UTR 3 NP_001278645.1
NM_001291717.1 2083 UTR 3 NP_001278646.1
NM_001291719.1 2083 UTR 3 NP_001278648.1
NM_001318126.1 2083 UTR 3 NP_001305055.1
NM_001318127.1 2083 UTR 3 NP_001305056.1
NM_006115.4 2083 UTR 3 NP_006106.1
NM_206953.2 2083 UTR 3 NP_996836.1
NM_206954.2 2083 UTR 3 NP_996837.1
NM_206955.2 2083 UTR 3 NP_996838.1
NM_206956.2 2083 UTR 3 NP_996839.1
XM_011530034.2 2083 UTR 3 XP_011528336.1

View Full Product Details