Product Details

SNP ID
rs114364590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44572382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGGCCGTGAAAACTCAAAAGA[C/T]ACAGTCTTTAAAATAAGGACAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00207 PubMed Links

Gene Details

Gene
LINC00207
Gene Name
long intergenic non-protein coding RNA 207
There are no transcripts associated with this gene.

View Full Product Details