Product Details

SNP ID
rs118009109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27753786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACATGGCAGGCCCTGGATTCTGG[C/T]AATACTGAGGCACAGTGCCCCAACC
Phenotype
MIM: 156100
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MN1 PubMed Links

Gene Details

Gene
MN1
Gene Name
MN1 proto-oncogene, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002430.2 Intron NP_002421.3

View Full Product Details