Product Details

SNP ID
rs114791146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27979126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCTGACTTGCTTTTCCTAGACT[A/G]GAGAAACAAAACGTTATTCCATTTT
Phenotype
MIM: 615098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC28 PubMed Links

Gene Details

Gene
TTC28
Gene Name
tetratricopeptide repeat domain 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145418.1 10683 UTR 3 NP_001138890.1
XM_005261405.2 10683 UTR 3 XP_005261462.1
XM_006724171.3 10683 UTR 3 XP_006724234.1
XM_011530018.2 10683 UTR 3 XP_011528320.1
XM_011530019.2 10683 Intron XP_011528321.1
XM_011530020.1 10683 Intron XP_011528322.1
XM_011530021.2 10683 Intron XP_011528323.1
XM_011530022.1 10683 Intron XP_011528324.1
XM_017028673.1 10683 UTR 3 XP_016884162.1
Gene
TTC28-AS1
Gene Name
TTC28 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details