Product Details

SNP ID
rs116730528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29329452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCACTGGTTCTGCAGGGTGCAG[C/T]TAGGAAGTGGGAACAATGGAGTTCC
Phenotype
MIM: 600157 MIM: 602220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP1B1 PubMed Links

Gene Details

Gene
AP1B1
Gene Name
adaptor related protein complex 1 beta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127.3 Intron NP_001118.3
NM_001166019.1 Intron NP_001159491.1
NM_145730.2 Intron NP_663782.2
Gene
MIR3653
Gene Name
microRNA 3653
There are no transcripts associated with this gene.

Gene
RASL10A
Gene Name
RAS like family 10 member A
There are no transcripts associated with this gene.

Gene
SNORD125
Gene Name
small nucleolar RNA, C/D box 125
There are no transcripts associated with this gene.

View Full Product Details