Product Details

SNP ID
rs116294680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50177786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCAACGAGAACCGCGACCACAT[A/C]AAGTCGCTCAACCGGCTGGACTTCA
Phenotype
MIM: 608421
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PANX2 PubMed Links

Gene Details

Gene
PANX2
Gene Name
pannexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160300.1 1074 Silent Mutation ATA,ATC I358I NP_001153772.1
NM_052839.3 1074 Silent Mutation ATA,ATC I358I NP_443071.2
Gene
TRABD
Gene Name
TraB domain containing
There are no transcripts associated with this gene.

View Full Product Details