Product Details

SNP ID
rs113054487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73583969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTTTGAGAGTTGCCTTTTTTCA[A/C]AGTAATCCAGGAGTATTGCCAAGGT
Phenotype
MIM: 300922
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHIC1 PubMed Links

Gene Details

Gene
CHIC1
Gene Name
cysteine rich hydrophobic domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039840.2 Intron NP_001034929.2
NM_001300884.1 Intron NP_001287813.1
XM_017029582.1 Intron XP_016885071.1
XM_017029583.1 Intron XP_016885072.1
XM_017029584.1 Intron XP_016885073.1

View Full Product Details