Product Details
- SNP ID
-
rs113107845
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:140503904 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCACGCGCGCTGAGAGTGCGAT[G/T]CGATGGCGGGCGGCGGCGAGCTGGG
- Phenotype
-
MIM: 313430
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SOX3
PubMed Links
Gene Details
- Gene
- SOX3
- Gene Name
- SRY-box 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005634.2 |
1157 |
Missense Mutation |
|
|
NP_005625.2 |
View Full Product Details