Product Details

SNP ID
rs115756523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38823419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAGTCAAAATTACCTAACACA[C/T]ATCTCAGAACTTAGTTAGAGCAGCC
Phenotype
MIM: 605403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLR6 PubMed Links

Gene Details

Gene
TLR6
Gene Name
toll like receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006068.4 6161 Intron NP_006059.2
XM_005262637.4 6161 UTR 3 XP_005262694.1
XM_011513612.2 6161 UTR 3 XP_011511914.1
XM_011513613.2 6161 UTR 3 XP_011511915.1
XM_011513614.2 6161 UTR 3 XP_011511916.1

View Full Product Details