Product Details

SNP ID
rs112893386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176684007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATGAAAATCCTGGCTCACAAGC[C/T]TTCTGGCGGTTCGTACATGGCCGTC
Phenotype
MIM: 601528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VEGFC PubMed Links

Gene Details

Gene
VEGFC
Gene Name
vascular endothelial growth factor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005429.4 1613 Silent Mutation AAA,AAG K393K NP_005420.1

View Full Product Details